Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CEACAM5/CD66e Antibody (161 (NCRC16)), Biotin, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP250161B
Description
CEACAM5/CD66e Monoclonal specifically detects CEACAM5/CD66e in Human samples. It is validated for Western Blot, ELISA.Specifications
CEACAM5/CD66e | |
Monoclonal | |
Biotin | |
PBS with 0.05% Sodium Azide | |
CEACAM5 | |
Colon cancer cell lines | |
0.1 mL | |
Primary | |
Human | |
Purified |
Western Blot, ELISA | |
161 (NCRC16) | |
Western Blot, ELISA | |
Carcinoembryonic antigen, carcinoembryonic antigen-related cell adhesion molecule 5, CD66e antigen, CEACD66e, DKFZp781M2392, Meconium antigen 100 | |
Mouse | |
Protein A purified | |
RUO | |
1048 | |
Store at 4C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction