Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CEACAM5/CD66e Antibody (Arcitumomab) - Azide and BSA Free, Novus Biologicals™

Human Monoclonal Antibody
Supplier: Novus Biologicals NBP2759040.2MG
Description
CEACAM5/CD66e Monoclonal specifically detects CEACAM5/CD66e in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunofluorescence.Specifications
CEACAM5/CD66e | |
Monoclonal | |
Unconjugated | |
PBS with 0.02% Proclin 300 | |
CEACAM5 | |
Carcinoembryonic antigen (CEA) describes a set of highly related glycoproteins involved in cell adhesion normally produced in gastrointestinal tissue during fetal development, but only at very low levels in adults. | |
Primary | |
Recognises human carcinoembryonic antigen. | |
Store at 4°C for up to 3 months. For longer storage, aliquot and store at -20°C. |
Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunofluorescence | |
Arcitumomab | |
Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunofluorescence | |
Carcinoembryonic antigen, carcinoembryonic antigen-related cell adhesion molecule 5, CD66e antigen, CEACD66e, DKFZp781M2392, Meconium antigen 100 | |
Human | |
0.2 mg | |
1048 | |
Human | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction