Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CEACAM5/CD66e Antibody (CB30), Alexa Fluor™ 488, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB11058734AF488
Description
CEACAM5/CD66e Monoclonal specifically detects CEACAM5/CD66e in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.Specifications
CEACAM5/CD66e | |
Monoclonal | |
Alexa Fluor 488 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
CEACAM5 | |
Human carcinoembryonic antigen (CEA; CEACAM5) | |
0.1 mL | |
Cancer, Cellular Markers, Immunology | |
1048 | |
Protein A purified | |
Store at 4C in the dark. | |
IgG1 |
Flow Cytometry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
CB30 | |
Flow Cytometry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen | |
Carcinoembryonic antigen, carcinoembryonic antigen-related cell adhesion molecule 5, CD66e antigen, CEACD66e, DKFZp781M2392, Meconium antigen 100 | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
The antibody CB30 recognizes CD66e (CEA; 180-200 kDa), a cell surface bound carcinoembryonic antigen mainly expressed on epithelial cells. | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction