Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CEACAM5/CD66e Antibody (SPM541), mFluor Violet 610 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP234787MFV610
Description
CEACAM5/CD66e Monoclonal antibody specifically detects CEACAM5/CD66e in Human, Monkey samples. It is validated for Flow CytometrySpecifications
CEACAM5/CD66e | |
Monoclonal | |
mFluor Violet 610 SE | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human, Monkey | |
Purified |
Flow Cytometry | |
SPM541 | |
Flow Cytometry | |
Carcinoembryonic antigen, carcinoembryonic antigen-related cell adhesion molecule 5, CD66e antigen, CEACD66e, DKFZp781M2392, Meconium antigen 100 | |
Human colon carcinoma extract (Uniprot: P06731) | |
0.1 mL | |
Apoptosis, Cancer, Cellular Markers, Immunology | |
1048 | |
Store at 4°C in the dark. | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction