Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CEACAM5 (CD66e) Mouse anti-Human, Clone: CB30, BD Pharmingen™
Mouse Monoclonal Antibody
Supplier: BD Biosciences 568709
This item is not returnable.
View return policy
Description
CEACAM5 (CD66e) Monoclonal antibody specifically detects CEACAM5 (CD66e) in Human samples. It is validated for Flow cytometry (Routinely Tested), Immunohistochemistry (Tested During Development)
Specifications
CEACAM5 (CD66e) | |
Monoclonal | |
0.5 mg/mL | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Mouse | |
Affinity Chromatography | |
RUO | |
Human | |
Purified |
Flow Cytometry, Immunohistochemistry | |
CB30 | |
Unconjugated | |
CD66e; CEACAM-5; CEACAM5; Carcinoembryonic antigen-related cell adhesion molecule 5 | |
CEACAM5 Protein | |
0.1 mg | |
Primary | |
Store undiluted at 4°C. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. | |
IgG1 κ |
Safety and Handling
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction