Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CEACAM5 (CD66e) Mouse anti-Human, Clone: M5A, MilliporeSigma™

Catalog No. MABC112325 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABC1123-25 25 μg
MABC1123-100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MABC1123-25 Supplier MilliporeSigma Supplier No. MABC112325UG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-CEACAM5 (CD66e), clone M5A is a mouse monoclonal antibody that detects Carcinoembryonic antigen-related cell adhesion molecule 5 and has been tested for use in Flow Cytometry, Immunohistochemistry, and Radioimmunoassay.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CEACAM5 (CD66e)
Applications Flow Cytometry, Immunohistochemistry, Radioimmune Assays (RIA)
Classification Monoclonal
Clone M5A
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Gene Symbols CEACAM5,CEA
Host Species Mouse
Immunogen Full length purified human Carcinoembryonic antigen-related cell adhesion molecule 5 (CEA).
Purification Method Protein A Purified
Quantity 25 μg
Regulatory Status RUO
Research Discipline Apoptosis and Cancer
Primary or Secondary Primary
Gene ID (Entrez) NP_001278413
Test Specificity Clone M5A is a mouse monoclonal antibody that detects human Carcinoembryonic antigen-related cell adhesion molecule 5 (CEA).
Target Species Human
Content And Storage Stable for 1 year at 2-8°C from date of receipt.
Form Purified
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.