Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CEACAM5, Mouse anti-Human, Clone: CB30, Abnova™

Catalog No. 89999044 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-999-044 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-999-044 Supplier Abnova Corporation Supplier No. MAB16246
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Specifications

Antigen CEACAM5
Applications Flow Cytometry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation
Classification Monoclonal
Clone CB30
Conjugate Unconjugated
Dilution Flow Cytometry Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (10 ug/mL) Immunohistochemistry (Frozen sections) Immunoprecipitation The optimal working dilution should be determined by the end user.
Formulation In PBS (0.09% sodium azide).
Gene carcinoembryonic antigen-related cell adhesion molecule 5
Gene Alias CD66e/CEA/DKFZp781M2392
Gene Symbols CEACAM5
Host Species Mouse
Immunogen Purified human carcinoembryonic antigen.
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1048
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.