Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CEACAM5 Rabbit anti-Human, Unconjugated, Clone: RM326, Abnova™

Catalog No. 89339187 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-339-187 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-339-187 Supplier Abnova Corporation Supplier No. MAB21970
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody.

CEACAM5 Monoclonal antibody specifically detects CEACAM5 in Human samples. It is validated for ELISA, WB-Ce, IHC-P.

Specifications

Antigen CEACAM5
Applications ELISA, Western Blot, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone RM326
Conjugate Unconjugated
Description carcinoembryonic antigen-related cell adhesion molecule 5
Gene CEACAM5
Gene Alias CD66e, CEA, DKFZp781M2392
Host Species Rabbit
Immunogen Native CEA protein purified from human liver tissue
Quantity 100 μL
Primary or Secondary Primary
Gene ID (Entrez) 1048
Target Species Human
Content And Storage Store at -20°C. Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.