Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CEACAM6/CD66c Antibody (1H7-4B), mFluor Violet 610 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP254626MFV610
Description
CEACAM6/CD66c Monoclonal antibody specifically detects CEACAM6/CD66c in Human, Mouse samples. It is validated for Flow CytometrySpecifications
CEACAM6/CD66c | |
Monoclonal | |
mFluor Violet 610 SE | |
50mM Sodium Borate | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Human, Mouse | |
Purified |
Flow Cytometry | |
1H7-4B | |
Flow Cytometry | |
carcinoembryonic antigen-related cell adhesion molecule 6, carcinoembryonic antigen-related cell adhesion molecule 6 (non-specific crossreacting antigen), CD66c, CD66c antigen, NCACEAL, Non-specific crossreacting antigen, Normal cross-reacting antigen | |
Recombinant human CEACAM6/CD66c extracellular domain. | |
0.1 mL | |
Cancer | |
4680 | |
Store at 4C in the dark. | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction