Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CEACAM6/CD66c Rabbit anti-Human, Clone: 408, Novus Biologicals™

Rabbit Monoclonal Antibody
$353.50 - $560.50
Specifications
Antigen | CEACAM6/CD66c |
---|---|
Clone | 408 |
Dilution | Western Blot 1:500-1:1000, Flow Cytometry 1:100-1:500, ELISA 1:25000-1:50000, Immunocytochemistry/Immunofluorescence 1:100-1:500 |
Applications | Western Blot, Flow Cytometry, ELISA, Immunofluorescence |
Classification | Monoclonal |
Description
CEACAM6/CD66c Monoclonal specifically detects CEACAM6/CD66c in Human samples. It is validated for Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence.Specifications
CEACAM6/CD66c | |
Western Blot 1:500-1:1000, Flow Cytometry 1:100-1:500, ELISA 1:25000-1:50000, Immunocytochemistry/Immunofluorescence 1:100-1:500 | |
Monoclonal | |
Purified | |
RUO | |
Human | |
carcinoembryonic antigen-related cell adhesion molecule 6, carcinoembryonic antigen-related cell adhesion molecule 6 (non-specific crossreacting antigen), CD66c, CD66c antigen, NCACEAL, Non-specific crossreacting antigen, Normal cross-reacting antigen | |
Recombinant Human CEACAM6/CD66c protein | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
408 | |
Western Blot, Flow Cytometry, ELISA, Immunofluorescence | |
Unconjugated | |
Rabbit | |
Cancer | |
0.2 um filtered solution in PBS | |
4680 | |
IgG | |
Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title