Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CEACAM6 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5102976
Description
Antibody detects endogenous levels of total CEACAM6.
CEA Cell Adhesion Molecule 6 (CEACAM6) belongs to the carcinoembryonic antigen (CEA) family whose members are glycosyl phosphatidyl inositol (GPI) anchored cell surface glycoproteins. Members of this family play a role in cell adhesion and are widely used as tumor markers in serum immunoassay determinations of carcinoma. CEACAM6 affects the sensitivity of tumor cells to adenovirus infection. The acts as a receptor for adherent-invasive E. coli adhesion to the surface of ileal epithelial cells in patients with Crohn's disease.
Specifications
CEACAM6 | |
Polyclonal | |
Unconjugated | |
CEACAM6 | |
BGP; BGP-1; carcinoembryonic antigen related cell adhesion molecule 6; carcinoembryonic antigen-related cell adhesion molecule 6; carcinoembryonic antigen-related cell adhesion molecule 6 (non-specific cross reacting antigen); CD66a; CD66c; CD66d; CD66e; CEA; CEACAM-1; CEACAM-3; CEACAM-5; CEACAM6; CEACAM-6; CEAL; CGM1; Cluster of Differentiation 66c; NCA; Non-specific crossreacting antigen; Normal cross-reacting antigen; pan CEACAM | |
Rabbit | |
Affinity chromatography | |
RUO | |
100125369, 4680 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
P40199 | |
CEACAM6 | |
A synthesized peptide derived from human CEACAM6(Accession P40199), corresponding to amino acid residues P220-S270. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction