Learn More
                                    CEACAM7 Antibody (962720), mFluor Violet 500 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB44782MFV500
Description
CEACAM7 Monoclonal antibody specifically detects CEACAM7 in Human samples. It is validated for Flow Cytometry, ELISA, Immunohistochemistry, CyTOFSpecifications
| CEACAM7 | |
| Monoclonal | |
| mFluor Violet 500 SE | |
| 50mM Sodium Borate | |
| Mouse | |
| 0.1 mL | |
| Cancer | |
| 1087 | |
| Store at 4C in the dark. | 
| Flow Cytometry, ELISA, Immunohistochemistry, CyTOF | |
| 962720 | |
| Flow Cytometry, ELISA, Immunohistochemistry, CyTOF-ready | |
| Carcinoembryonic antigen CGM 2, Carcinoembryonic antigen CGM2, Carcinoembryonic antigen gene family member 2, Carcinoembryonic antigen related cell adhesion molecule 7, CEA, CEACAM 7, CGM 2, CGM2 | |
| Chinese Hamster Ovary cell line, CHO-derived recombinant human CEACAM-7, Thr36-Asn233, Accession # Q14002 | |
| RUO | |
| Primary | |
| Human | |
| IgG1 | 