Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CEACAM7 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA576345
Description
The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).
CEACAM7 is a CEA-related cell adhesion molecule 7 that belongs to the carcinoembryonic antigen (CEA) family. CEACAM7 encodes a glycosyl phosphatidyl inositol (GPI)-linked glycoprotein which is only found on the apical surface of highly differentiated epithelial cells adult colon and on a small epithelial cell population in pancreatic ducts. CEACAM7 expression is completely lost upon malignant transformation already in hyperplastic polyps and early adenomas as well as in adenocarcinomas of the colon. Like all members of the CEACAM family, it consists of a single N domain, with structural homology to the immunoglobulin variable domains, followed by one immunoglobulin constant-like A domain.
Specifications
CEACAM7 | |
Polyclonal | |
Unconjugated | |
CEACAM7 | |
BGP; BGP7; carcinoembryonic antigen CGM2; carcinoembryonic antigen gene family member 2; carcinoembryonic antigen related cell adhesion molecule 7; carcinoembryonic antigen-related cell adhesion molecule 7; CEA; CEACAM7; CGM2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
1087 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
Q14002 | |
CEACAM7 | |
Recombinant fusion protein of human CEACAM7. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction