Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CEACAM8/CD66b Antibody (013), DyLight 350 , Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP289971UV
Description
CEACAM8/CD66b Monoclonal antibody specifically detects CEACAM8/CD66b in Human samples. It is validated for ELISASpecifications
CEACAM8/CD66b | |
Monoclonal | |
DyLight 350 | |
Carcinoembryonic antigen CGM6, carcinoembryonic antigen gene family member 6, carcinoembryonic antigen-related cell adhesion molecule 8, CD66b, CD66b antigen, CD67, CD67 antigen, CGM6, NCA-95, Non-specific cross-reacting antigen NCA-95 | |
This antibody was obtained from a rabbit immunized with purified, recombinant Human CEACAM8/CD66b (NP_001807.2; Met 1-Ser 319). | |
0.1 mL | |
Cancer | |
1088 | |
Store at 4°C in the dark. | |
IgG |
ELISA | |
013 | |
50mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction