Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CEACAM8/CD66b Antibody (6/40c), Janelia Fluor™ 549, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP254627JF549
Description
CEACAM8/CD66b Monoclonal specifically detects CEACAM8/CD66b in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry.Specifications
CEACAM8/CD66b | |
Monoclonal | |
Janelia Fluor 549 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
CEACAM8 | |
Recombinant soluble human CEACAM8-Fc produced in HEK293 cells | |
0.1 mL | |
Primary | |
Clone 6/40c does not cross-react with human CEACAM1/3/4/5/6/7, rat CECAM1, or mouse CEACAM1/2. | |
Store at 4°C in the dark. | |
IgG1 κ |
Western Blot, Flow Cytometry, ELISA, Immunohistochemistry | |
6/40c | |
Western Blot, Flow Cytometry, ELISA, Immunohistochemistry | |
Carcinoembryonic antigen CGM6, carcinoembryonic antigen gene family member 6, carcinoembryonic antigen-related cell adhesion molecule 8, CD66b, CD66b antigen, CD67, CD67 antigen, CGM6, NCA-95, Non-specific cross-reacting antigen NCA-95 | |
Mouse | |
Protein A purified | |
Cancer | |
1088 | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction