Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CEACAM8/CD66b Antibody (80H3), Novus Biologicals™

Mouse Monoclonal Antibody
$207.00 - $499.50
Specifications
Antigen | CEACAM8/CD66b |
---|---|
Clone | 80H3 |
Concentration | 1.0 mg/mL |
Dilution | Western Blot 1:100-1:2000, Flow Cytometry 1:50-1:100, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Frozen 1:25-1:100 |
Applications | Western Blot, Immunohistochemistry (Frozen), Immunohistochemistry, Flow Cytometry |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB017664
![]() |
Novus Biologicals
NB100649160.025MG |
0.025 mg |
Each for $207.00
|
|
|||||
NB10064916
![]() |
Novus Biologicals
NB10064916 |
0.1 mg |
Each for $499.50
|
|
|||||
Description
CEACAM8/CD66b Monoclonal specifically detects CEACAM8/CD66b in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen.Specifications
CEACAM8/CD66b | |
1.0 mg/mL | |
Western Blot, Immunohistochemistry (Frozen), Immunohistochemistry, Flow Cytometry | |
Unconjugated | |
Mouse | |
Human | |
Carcinoembryonic antigen CGM6, carcinoembryonic antigen gene family member 6, carcinoembryonic antigen-related cell adhesion molecule 8, CD66b, CD66b antigen, CD67, CD67 antigen, CGM6, NCA-95, Non-specific cross-reacting antigen NCA-95 | |
CEACAM8 | |
IgG1 | |
Protein A purified | |
NB100-64916 reacts with the CGM6 gene product of the carcinoembryonic gene family. This is a 95-100kD GPI anchored molecule. CEACAM8 is expressed strongly by mature granulocytes and metamyelocytes but weakly by bone marrow myelocytes. CEACAM8 is not expressed by peripheral blood monocytes or lymphocytes. |
80H3 | |
Western Blot 1:100-1:2000, Flow Cytometry 1:50-1:100, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Frozen 1:25-1:100 | |
Monoclonal | |
Purified | |
RUO | |
P31997 | |
1088 | |
CML leucocytes | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title