Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CEACAM8/CD66b Antibody (913542), mFluor Violet 610 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB4246MFV610
Description
CEACAM8/CD66b Monoclonal antibody specifically detects CEACAM8/CD66b in Human samples. It is validated for Western Blot, Flow Cytometry, CyTOFSpecifications
| CEACAM8/CD66b | |
| Monoclonal | |
| mFluor Violet 610 SE | |
| 50mM Sodium Borate | |
| Mouse | |
| 0.1 mL | |
| Cancer | |
| 1088 | |
| Store at 4C in the dark. |
| Western Blot, Flow Cytometry, CyTOF | |
| 913542 | |
| Western Blot, Flow Cytometry, CyTOF-ready | |
| Carcinoembryonic antigen CGM6, carcinoembryonic antigen gene family member 6, carcinoembryonic antigen-related cell adhesion molecule 8, CD66b, CD66b antigen, CD67, CD67 antigen, CGM6, NCA-95, Non-specific cross-reacting antigen NCA-95 | |
| E.coli-derived recombinant human CEACAM-8/CD66b, Gln35-His141, Accession # P31997 | |
| RUO | |
| Primary | |
| Human | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction