Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CEBP Beta Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$382.00 - $610.00
Specifications
Antigen | CEBP Beta |
---|---|
Applications | Western Blot, Immunocytochemistry, Immunofluorescence |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Description
CEBP Beta Polyclonal specifically detects CEBP Beta in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
CEBP Beta | |
Polyclonal | |
Rabbit | |
Human | |
C/EBP beta, C/EBP-beta, CCAAT/enhancer binding protein (C/EBP), beta, CCAAT/enhancer-binding protein beta, CRP2, IL6DBP, interleukin 6-dependent DNA-binding protein, LAPTCF-5, Liver activator protein, liver-enriched transcriptional activator protein, MGC32080, NF-IL6, Nuclear factor NF-IL6, nuclear factor of interleukin 6, TCF5NFIL6, Transcription factor 5 | |
CEBPB | |
IgG | |
Affinity Purified |
Western Blot, Immunocytochemistry, Immunofluorescence | |
Unconjugated | |
RUO | |
PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide | |
1051 | |
This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:GFPYALRAYLGYQAVPSGSSGSLSTS | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title