Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CEBP Delta Antibody, FITC, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB11085519F
Description
CEBP Delta Polyclonal antibody specifically detects CEBP Delta in Human, Mouse samples. It is validated for ELISA, Immunohistochemistry, KnockDownSpecifications
CEBP Delta | |
Polyclonal | |
ELISA, Immunohistochemistry, Knockdown Validated | |
C/EBP-delta, CCAAT/enhancer binding protein (C/EBP), delta, CCAAT/enhancer-binding protein delta, CELF, CRP3, NF-IL6-betaC/EBP delta, Nuclear factor NF-IL6-beta | |
This affinity purified CEBP Delta Antibody was prepared from whole rabbit serum produced by repeated immunizations with a recombinant protein corresponding to the amino terminus of mouse CEBP Delta. (Uniprot: Q00322) | |
0.1 mL | |
Primary | |
Human, Mouse | |
Purified |
ELISA, Immunohistochemistry, KnockDown | |
FITC | |
PBS | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
1052 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction