Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CEBP delta Polyclonal Antibody, Rockland™
SDP

Catalog No. 501991940
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
25 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-199-1940 100 μg
50-199-1941 25 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-199-1940 Supplier Rockland Immunochemicals Supplier No. 600401A61
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Store vial at -20°C prior to opening. Aliquot contents and freeze at -20°C or below for extended storage. Avoid cycles of freezing and thawing. Centrifuge product if not completely clear after standing at room temperature. This product is stable for several weeks at 4°C as an undiluted liquid. Dilute only prior to immediate use. This antibody recognizes endogenous mouse and recombinant human C/EBP delta protein. A BLAST analysis was used to suggest cross-reactivity with C/EBPd from human sources based on 77% homology with the immunizing sequence. No cross-reactivity is observed with mouse C/EBPa or C/EBPb. Reactivity with C/EBPd from other sources has not been determined.

C/EBP delta are CCAAT enhancer binding proteins (CEBPs), a family of transcription factors that all contain a highly conserved, basic-leucine zipper domain at the C-terminus that is involved in dimerization and DNA binding. At least six members of the family have been isolated and characterized to date (CEBP alpha to CEBP zeta). CEBPD is a leucine zipper (LZ) DNA-binding protein that regulates gene expression in a variety of tissues including liver, adipose, lung and intestine. CEBPD is an important transcriptional activator in the regulation of genes involved in immune and inflammatory responses and has been reported to possess many tumor suppressor-like properties.

Specifications

Antigen CEBP delta
Applications ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 0.02M potassium phosphate with 0.15M NaCl and 0.01% sodium azide; pH 7.2
Gene CEBPD
Gene Accession No. P49716, Q00322
Gene Alias C/EBP delta; C/EBPd; C/EBPdelta; C/EBP-delta; C/EBP-related protein 3; CCAAT/enhancer binding protein (C/EBP), delta; CCAAT/enhancer binding protein delta; CCAAT/enhancer-binding protein delta; Cebpd; Celf; Crp3; NF-IL6-beta; Nuclear factor NF-IL6-beta; Transcription factor CELF
Gene Symbols CEBPD
Host Species Rabbit
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1052, 12609
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.