Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CEBP gamma Antibody (001), DyLight 755, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP290304IR
Description
CEBP gamma Monoclonal specifically detects CEBP gamma in Human samples. It is validated for ELISA.Specifications
CEBP gamma | |
Monoclonal | |
DyLight 755 | |
50mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
ELISA | |
1 | |
ELISA | |
c/EBP gamma, CCAAT/enhancer binding protein (C/EBP), gamma, CCAAT/enhancer-binding protein gamma, GPE1BP, IG/EBP-1 | |
This antibody was obtained from a rabbit immunized with purified, recombinant Human CEBP gamma (P53567; Pro39-Asn147). | |
0.1 mL | |
Cell Cycle and Replication | |
1054 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction