Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CEBPA, Rabbit anti-Human, Polyclonal Antibody, Abnova™

Catalog No. 89999268 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-999-268 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-999-268 Supplier Abnova Corporation Supplier No. PAB31652
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Specifications

Antigen CEBPA
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution ELISA (1:10000) Western Blot (1:500-2000) The optimal working dilution should be determined by the end user.
Formulation In PBS (50% glycerol, 0.5% BSA and 0.02% sodium azide).
Gene CCAAT/enhancer binding protein (C/EBP), alpha
Gene Alias C/EBP-alpha/CEBP
Gene Symbols CEBPA
Host Species Rabbit
Immunogen A synthetic peptide corresponding to amino acids 170-250 of human CEBPA (phospho T226).
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1050
Test Specificity This antibody detects endogenous levels of C/EBP alpha protein only when phosphorylated at T226.
Target Species Human, Monkey, Mouse, Rat
Content And Storage Store at -20°C. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.