Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CEBPB Rabbit anti-Human, Unconjugated, Clone: AAG-3, Abnova™
Rabbit Monoclonal Antibody.
Supplier: Abnova Corporation MAB19888
Description
CEBPB Monoclonal antibody specifically detects CEBPB in Human samples. It is validated for IP, WB-Ce, Flow Cyt.Specifications
CEBPB | |
Monoclonal | |
Unconjugated | |
CEBPB | |
Rabbit | |
100 μg | |
1051 | |
Store at -20°C for one year. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for a longer time. Avoid repeated freezing and thawing. | |
IgG |
Immunoprecipitation, Western Blot, Flow Cytometry | |
AAG-3 | |
CCAAT/enhancer binding protein (C/EBP), beta | |
C/EBP-beta, CRP2, IL6DBP, LAP, MGC32080, NF-IL6, TCF5 | |
A synthetic peptide corresponding to human CEBPB | |
Primary | |
Human | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction