Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CECR1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA543014
Description
Peptide sequence: MQFRFAHPTP RPSEKCSKWI LLEDYRKRVQ NVTEFDDSLL RNFTLVTQHP Sequence homology: Human: 100%.
This gene encodes a member of a subfamily of the adenosine deaminase protein family. The encoded protein may act as a growth factor and have adenosine deaminase activity. It may be responsible for some of the phenotypic features associated with cat eye syndrome. Two transcript variants encoding distinct isoforms have been identified for this gene.
Specifications
CECR1 | |
Polyclonal | |
Unconjugated | |
ADA2 | |
ADA2; Adenosine deaminase 2; adenosine deaminase CECR1; ADGF; cat eye syndrome chromosome region, candidate 1; cat eye syndrome critical region protein 1; CECR1; IDGFL; PAN; SNEDS | |
Rabbit | |
Affinity Chromatography | |
RUO | |
51816 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q9NZK5 | |
ADA2 | |
Synthetic peptide directed towards the N-terminal of human CECR1 (aa 144-193). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction