Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CECR2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA582045
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA582045 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA582045 Supplier Invitrogen™ Supplier No. PA582045
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: TLGHVMDSRV MRPPVPPNQW TEQSGFLPHG VPSSGYMRPP CKSAGHRLQP PPVPAPSSLF GAPAQALRGV QGGDSMMDSP EMIAMQQLSS RVCPPGVPYH PHQPAHPRLP GPFPQVAHPM SVTVSAPKPA LGNPGRAPEN SEAQEP Highest antigen sequence indentity to the following orthologs: Rat - 79%, Mouse - 79%.

This gene encodes a bromodomain-containing protein that is involved in chromatin remodeling, and may additionally play a role in DNA damage response. The encoded protein functions as part of an ATP-dependent complex that is involved in neurulation. This gene is a candidate gene for Cat Eye Syndrome. Alternative splicing results in multiple transcript variants. [provided by RefSeq, Feb 2014].
TRUSTED_SUSTAINABILITY

Specifications

Antigen CECR2
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CECR2
Gene Accession No. Q9BXF3
Gene Alias cat eye syndrome chromosome region, candidate 2; cat eye syndrome critical region protein 2; CECR2; CECR2, histone acetyl-lysine reader; KIAA1740
Gene Symbols CECR2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human CECR2. Recombinant protein control fragment (Product #RP-91489).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 27443
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.