Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CECR5 Antibody (SR1791), Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP32258650UL
Description
CECR5 Monoclonal antibody specifically detects CECR5 in Human samples. It is validated for Western Blot, ImmunofluorescenceSpecifications
CECR5 | |
Monoclonal | |
Unconjugated | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot, Immunofluorescence | |
SR1791 | |
Western Blot 1:500-1:2000, Immunocytochemistry/Immunofluorescence 1:50-1:200 | |
cat eye syndrome chromosome region, candidate 5 | |
A synthesized peptide derived from human CECR5 (Uniprot #: Q9BXW7) | |
50 μg | |
Stem Cell Markers | |
27440 | |
Store at -20° C. Avoid freeze/thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction