Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CECR5 Recombinant Rabbit Monoclonal Antibody (JE64-76)
GREENER_CHOICE

Catalog No. PIMA544688
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA544688 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIMA544688 Supplier Invitrogen™ Supplier No. MA544688
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

CECR5 Recombinant Monoclonal Antibody for Western Blot, IHC (P)

HDHD5 (Haloacid Dehalogenase Like Hydrolase Domain Containing 5) is a Protein Coding gene. Diseases associated with HDHD5 include Cat Eye Syndrome. An important paralog of this gene is CRLS1. [GeneCards].
TRUSTED_SUSTAINABILITY

Specifications

Antigen CECR5
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Recombinant Monoclonal
Clone JE64-76
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.05% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4
Gene HDHD5
Gene Accession No. Q9BXW7
Gene Alias A930002G03Rik; cat eye syndrome chromosome region, candidate 5; cat eye syndrome chromosome region, candidate 5 homolog; cat eye syndrome critical region protein 5; Cat eye syndrome critical region protein 5 homolog; CECR5; Haloacid dehalogenase-like hydrolase domain-containing 5; HDHD5
Gene Symbols HDHD5
Host Species Rabbit
Immunogen Synthetic peptide within human CECR5 aa 351-400/423.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 27440
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.