Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CECR6 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA567013
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA567013 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA567013 Supplier Invitrogen™ Supplier No. PA567013
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PPPPPQGGSQL GHCISENEGG AHGYVNTLAV ASQN Highest antigen sequence identity to the following orthologs - mouse 86%, rat 86%.

CECR6 is a protein associated with Cat Eye Syndrome (CES), a developmental disorder associated with the duplication of a 2 Mb region of 22q11.2. Duplication usually takes in the form of a surpernumerary bisatellited isodicentric chromosome, resulting in four copies of the region (represents an inv dup(22)(q11)). CES is characterized clinically by the combination of coloboma of the iris and anal atresia with fistula, down-slanting palpebral fissures, preauricular tags and/or pits, frequent occurrence of heart and renal malformations, and normal or near-normal mental development.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CECR6
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene TMEM121B
Gene Accession No. Q9BXQ6
Gene Alias AW048664; cat eye syndrome chromosome region, candidate 6; cat eye syndrome chromosome region, candidate 6 homolog; cat eye syndrome critical region protein 6; Cat eye syndrome critical region protein 6 homolog; Cecr6; RGD1565302; TMEM121B; Transmembrane protein 121B
Gene Symbols TMEM121B
Host Species Rabbit
Immunogen Recombinant Human CECR6. Recombinant protein control fragment (Product #RP-107653).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 27439
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.