Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cecropin BRabbit anti-Insect, Polyclonal, Rockland™
SDP

Catalog No. 501990053
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-199-0053 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-199-0053 Supplier Rockland Immunochemicals Supplier No. 100401242
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Store vial at -20°C prior to opening. Aliquot contents and freeze at -20°C or below for extended storage. Avoid cycles of freezing and thawing. Centrifuge product if not completely clear after standing at room temperature. This product is stable for several weeks at 4°C as an undiluted liquid. Dilute only prior to immediate use. Assay by immunoelectrophoresis resulted in a single precipitin arc against anti-Rabbit Serum and purified and partially purified cecropin B. Reactivity against other forms of cecropins has not yet been determined.

Cecropins are produced by insects, particularly under conditions of infection. Cecropins are bioactive peptides that exhibit activities by interacting with membranes and forming transmembrane channels that allow the free flow of electrolytes, metabolites and water across the phospholipid bilayers. Cecropins A, B and D are close homologues consisting of 35-39 residues. They are found in the pupae of the cecropin moth, but related homologues named lepodopteran, bactericidin, moricin and sarcotoxin are produced by other insects.

Specifications

Antigen Cecropin B
Applications ELISA, Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.33 mg/mL
Conjugate Unconjugated
Formulation 0.02M potassium phosphate with 0.15M NaCl, 50% glycerol and 0.01% sodium azide; pH 7.2
Host Species Rabbit
Immunogen This affinity purified antibody was prepared from whole rabbit serum produced by repeated immunizations with a synthetic full-length cecropin B corresponding to a modified sequence from that which is found in silkworm: KWKVFKKIEK VGRNIRNGIV KAGPAIAVL GEAKAL.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Insect
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.