Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cecropin Mouse anti-Human, Clone: 1D8, Bioss
SDP

Catalog No. 501979907
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-197-9907 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-197-9907 Supplier Bioss Supplier No. BSM2079M
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Cecropins are antimicrobial peptides.They were first isolated from the hemolymph of Hyalophora cecropia, from whence the term cecropin was derived. Cecropins lyse bacterial cell membranes; they also inhibit proline uptake and cause leaky membranes. Cecropins constitute a main part of the cell-free immunity of insects. Cecropins are small proteins of about 31 - 37 amino acid residues active against both Gram-positive and Gram-negative bacteria. Cecropins isolated from insects other than Hyalophora cecropia (Cecropia moth) have been given various names; bactericidin, lepidopteran, sarcotoxin, etc. All of these peptides are structurally related. Cecropin P1, an intestinal antibacterial peptide from Sus scrofa (Pig), also belongs to this family. Cecropin family also consists Cecropin A and Cecropin B.

Specifications

Antigen Cecropin
Applications Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 1D8
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Host Species Mouse
Immunogen KLH conjugated synthetic peptide derived from Hyalophora cecropia.
Purification Method Protein G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.