Learn More
Thermo Scientific™ CEDIA Mitragynine (Kratom) 200ng/mL Calibrator
Catalog No. 09980606
This item is not returnable. View return policy
For Criminal Justice and Forensic Use Only.
Catalog No. 09-980-606
Supplier: Thermo Scientific™ 10026593/DEL
This item is not returnable. View return policy
For Criminal Justice and Forensic Use Only.
Validate the CEDIA Mitragynine (Kratom) Assay using the CEDIA Mitragynine (Kratom) Drugs of Abuse Calibrators and Controls (CJ&F only).
Validate the Thermo Scientific™ CEDIA™ Mitragynine (Kratom) Assay and ensure accuracy of results using the Thermo Scientific CEDIA Mitragynine (Kratom) Calibrators and Controls (CJ&F). The five calibrators (including negative) establish a standard curve that correlates the instrument signals with the calibrators' respective concentrations in ng/mL. This allows for semi-quantitative measurements. Two controls, one above and one below the cutoff (50ng/mL), validate the calibration curve. The cutoff calibrator provides a reference point for qualitative detection of positive or negative results. For Criminal Justice and Forensic use only in the USA.
  • Ensures assay accuracy
  • Supports qualitative and semi-quantitative measurements
  • Available April 2019
TRUSTED_SUSTAINABILITY
For Criminal Justice and Forensic Use Only.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.