Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ CEDIA™ THC Multi-Level Control Set

Catalog No. p-4527682
Click to view available options
Concentration:
100 ng/mL
25 ng/mL
50 ng/mL
This item is not returnable. View return policy

Catalog No. 23-278-283

Supplier: Thermo Scientific™ 1661086

Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Eliminate time-consuming steps yet achieve high analytical accuracy with the ready-to-use Thermo Scientific™ CEDIA™ THC (Cannabinoid) Multi-Level Control Set.

Eliminate time-consuming steps yet achieve high analytical accuracy with the ready-to-use Thermo Scientific™ CEDIA™ THC (Cannabinoid) Drugs of Abuse Control Set in liquid form. These assays have been field-tested on a wide range of analyzers, and they are enthusiastically accepted in laboratories where cost control and optimal productivity are key issues.

  • Largest menu of tests available
  • Qualitative or semi-quantitative options for results
  • Excellent correlation to GC/MS
  • Applications for an array of clinical chemistry analyzers
  • Variety of calibrators and multiconstituent control sets
TRUSTED_SUSTAINABILITY

Specifications

Description CEDIA THC 25 Multi-Level Control Set
Concentration 25 ng/mL
Detectable Analytes 11-nor-9-COOH-D9-THC
Sample Type Urine
Type Control
Level Low (18.75ng/mL) and High (31.25ng/mL)
Quantity 1 x 15 mL Low and 1 x 15 mL High
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.