Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ CEDIA™ Therapeutic Drug Monitoring (TDM) Assays

Catalog No. 23278235
Click to view available options
Description:
Carbamazepine Assay
Gentamicin II Assay
Phenobarbital II Assay
Phenytoin II Assay
Procainamide Assay
Theophylline II Assay
Valproic Acid Assay

Catalog No. 23-278-235

Supplier: Thermo Scientific™ 100008

Only null left
Add to Cart
Add to Cart

Receive reliable performance on a wide range of analyzers with Thermo Scientific™ CEDIA™ Therapeutic Drug Monitoring (TDM) Assays.

Receive reliable performance on a wide range of analyzers with Thermo Scientific™ CEDIA™ Therapeutic Drug Monitoring (TDM) Assays. Manufactured since 1986, they provide an excellent range of Thermo Scientific™ DRI™ TDM Assays as well as CEDIA TDM Assays and Calibrators.

  • Wide dynamic measuring range of analytes as low as 0.15ng/mL for Digoxin
  • Extended linearity for analytes like Cyclosprine at 25ng/mL to 2000ng/mL range
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 2°C to 8°C
Description Theophylline II Assay
Sample Type Plasma, Serum
Type Reagent
Quantity 17 x 17 mL
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.