Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Thermo Scientific™ CEDIA™ Therapeutic Drug Monitoring (TDM) Assays
Click to view available options
Description:
Carbamazepine Assay
Gentamicin II Assay
Phenobarbital II Assay
Phenytoin II Assay
Procainamide Assay
Theophylline II Assay
Valproic Acid Assay
Description
Receive reliable performance on a wide range of analyzers with Thermo Scientific™ CEDIA™ Therapeutic Drug Monitoring (TDM) Assays. Manufactured since 1986, they provide an excellent range of Thermo Scientific™ DRI™ TDM Assays as well as CEDIA TDM Assays and Calibrators.
- Wide dynamic measuring range of analytes as low as 0.15ng/mL for Digoxin
- Extended linearity for analytes like Cyclosprine at 25ng/mL to 2000ng/mL range

Specifications
Specifications
Content And Storage | 2°C to 8°C |
Description | Carbamazepine Assay |
Detectable Analytes | Carbamazepine |
Sample Type | Plasma, Serum |
Type | Reagent |
Quantity | 17 x 17 mL |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction