Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cefadroxil, MilliporeSigma™ Supelco™
Shop All MilliporeSigma Supelco Products
Click to view available options
Quantity:
1 g
5 g
Description
Featured Industries: Pharmaceutical (small molecule)
Specifications
Specifications
| CAS | 50370-12-2 |
| Physical Form | Powder/Crystals |
| Linear Formula | C16H17N3O5 S |
| MDL Number | MFCD00865091 |
| Type | Analytical Standard |
| Quantity | 5 g |
| Formula Weight | 363.39 |
| Grade | Analytical Standard |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction