Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Cefazolin sodium salt, 98%

Catalog No. ac455210050
Click to view available options
Quantity:
5 g

Cefazolin sodium, CAS # 27164-46-1, is the sodium salt of the beta-lactam antibiotic cefazolin, a bactericidal compound of the cephalosporin family.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

• Cefazolin sodium is a beta-lactam antibiotic of the cephalosporin family

Applications

• Cefazolin binds and inactivates penicillin-binding proteins (PBP) and interferes with peptidoglycan chain crosslinking

• PBP inactivation leads to destruction of the cell wall and lysis

• Cefazolin exhibits broad spectrum activity against gram-positive and gram-negative bacteria

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 27164-46-1
Molecular Formula C14H16N8NaO4S3
Molecular Weight (g/mol) 479.504
MDL Number MFCD0005688
InChI Key MTIAAUXSENDLGW-SLNAEPSVSA-N
PubChem CID 131673922
IUPAC Name (6R,7R)-3-[(5-methyl-1,3,4-thiadiazol-2-yl)sulfanylmethyl]-8-oxo-7-[[2-(tetrazol-1-yl)acetyl]amino]-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid;molecular hydrogen;sodium
SMILES [HH].CC1=NN=C(S1)SCC2=C(N3C(C(C3=O)NC(=O)CN4C=NN=N4)SC2)C(=O)O.[Na]

Specifications

Type Cefazolin Sodium Salt
Melting Point 170°C to 177°C
Odor Odorless
Color White to Yellow
Quantity 5 g
Formula Weight 476.48
Percent Purity 98%
Chemical Name or Material Cefazolin sodium salt
Hazard Category Danger
Hazard Statement GHS H Statement
H334-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause an allergic skin reaction.
Precautionary Statement GHS P Statement
P284a-P261-P280-P342+P311-P363-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.
Wash contaminated clothing before reuse.
Dispose of contents/container in accordance with local/regional/national/international regulations.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.