Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cefotetan Disodium, G-Biosciences
SCI_ED

Catalog No. S902843
Click to view available options
Quantity:
100 mg
1 g

Cefotetan disodium is a semi-synthetic Beta-lactam antibiotic that is classified as a second generation cephalosporin.

Cefotetan disodium effects a broad range of gram positive and gram negative bacteria. It has also displayed a significant degree of bactericidal activity towards aerobic and anaerobic bacteria.

Chemical Identifiers

Molecular Weight (g/mol) 619.58 g/mol

Specifications

Boiling Point 798°C
Quantity 100 mg
Percent Purity >95%
Grade Laboratory
Physical Form Powder
Product Line Gibco
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.