Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cefotetan disodium, Thermo Scientific Chemicals
Click to view available options
Quantity:
100 mg
1 g
Description
A broad spectrum antibiotic
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Type | Cefotetan diSodium |
| Physical Form | Powder |
| Color | White |
| Quantity | 1 g |
| Boiling Point | 798°C |
| Solubility Information | Freely soluble in aqueous solution |
| Formula Weight | 619.58 |
| Chemical Name or Material | Cefotetan disodium |
Safety and Handling
| Hazard Category | H315-H317-H319-H334 |
| Hazard Statement | GHS H Statement H334-H317 May cause allergy or asthma symptoms or breathing difficulties if inhaled. May cause an allergic skin reaction. |
| Precautionary Statement | P261-P264b-P272-P280-P285-P302+P352-P304+P341-P305+P351+P338-P333+P313-P342+P311-P362-P501c |
| EINECSNumber | 277-834-9 |
| RTECSNumber | XI0330600 |
| TSCA | No |
| storageNote1 | 2° to 8°C |
| Recommended Storage | Keep cold |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction