Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cefpirome sulfate, 95%, Thermo Scientific Chemicals

Catalog No. AAJ6600106
Click to view available options
Quantity:
1 g
5 g

A fourth generation cephalosporin antibiotic

A fourth generation cephalosporin antibiotic

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 98753-19-6
Molecular Formula C22H24N6O9S3
Molecular Weight (g/mol) 612.647
MDL Number MFCD11864966
InChI Key RKTNPKZEPLCLSF-PXPMWPIZSA-N
PubChem CID 134129654
IUPAC Name (6S,7R)-7-[[2-(2-amino-1,3-thiazol-4-yl)-2-methoxyiminoacetyl]amino]-3-(6,7-dihydro-5H-cyclopenta[b]pyridin-1-ium-1-ylmethyl)-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid;hydrogen sulfate
SMILES CON=C(C1=CSC(=N1)N)C(=O)NC2C3N(C2=O)C(=C(CS3)C[N+]4=CC=CC5=C4CCC5)C(=O)O.OS(=O)(=O)[O-]

Specifications

Type Cefpirome Sulfate
Physical Form Powder
Melting Point 198°C to 202°C
Color White
Quantity 5 g
Solubility Information Soluble in DMSO
Formula Weight 612.66
Percent Purity 95%
Chemical Name or Material Cefpirome sulfate
RTECSNumber UW8970000
TSCA No
storageNote1 2° to 8°C
Recommended Storage Keep cold

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.