Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Cefprozil

Click to view available options
Quantity:
100mg
1g
92665-29-7
C18H19N3O5S
389.426
MFCD00865082
WDLWHQDACQUCJR-JQAPWBFTSA-N
123134496
(6S,7S)-7-[[(2S)-2-amino-2-(4-hydroxyphenyl)acetyl]amino]-8-oxo-3-[(E)-prop-1-enyl]-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid
CC=CC1=C(N2C(C(C2=O)NC(=O)C(C3=CC=C(C=C3)O)N)SC1)C(=O)O
92665-29-7
C18H19N3O5S
389.426
MFCD00865082
WDLWHQDACQUCJR-JQAPWBFTSA-N
123134496
(6S,7S)-7-[[(2S)-2-amino-2-(4-hydroxyphenyl)acetyl]amino]-8-oxo-3-[(E)-prop-1-enyl]-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid
CC=CC1=C(N2C(C(C2=O)NC(=O)C(C3=CC=C(C=C3)O)N)SC1)C(=O)O
Merck Index 14,1942
Solubility Information Soluble in DMSO at 10mg/mL; Insoluble in water
Formula Weight 389.42
Physical Form Powder
Boiling Point 803°C
Color White
Melting Point 218°C to 225°C
Quantity 100mg
Chemical Name or Material Cefprozil

RTECSNumber : XI0339000

storageNote1 : 2° to 8°C