Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ceftiofur Sodium, 95%, PROMO, Thermo Scientific Chemicals

Catalog No. J6688606PM
Click to view available options
Quantity:
5 g

A broad spectrum antibiotic

Applications

Ceftiofur sodium is resistant to the antibiotic resistance enzyme beta-lactamase, and has activity against Gram-positive and Gram-negative bacteria. E. coli strains resistant to ceftiofur have been reported. The bactericidal activity of ceftiofur results from the inhibition of cell wall synthesis via affinity for penicillin-binding proteins. The PBPs are transpeptidases which are vital in peptidoglycan biosynthesis. Therefore, their inhibition prevents this vital cell wall component from being properly synthesized.

Solubility

Freely soluble in aqueous solution

Notes

Research Use only: Not intended for animal or human diagnostic or therapeutic use.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 104010-37-9
Molecular Formula C19H16N5NaO7S3
Molecular Weight (g/mol) 545.53 g/mol
MDL Number MFCD01766184
InChI Key RFLHUYUQCKHUKS-JUODUXDSSA-M
PubChem CID 131632581
IUPAC Name sodium (6R,7R)-7-[(2Z)-2-(2-amino-1,3-thiazol-4-yl)-2-(methoxyimino)acetamido]-3-[(furan-2-carbonylsulfanyl)methyl]-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylate
SMILES [Na+].CO\N=C(/C(=O)N[C@H]1[C@H]2SCC(CSC(=O)C3=CC=CO3)=C(N2C1=O)C([O-])=O)C1=CSC(N)=N1

Specifications

Type Ceftiofur Sodium
Physical Form Powder
Color White or Yellow
Quantity 5 g
Solubility Information Freely soluble in aqueous solution
Percent Purity 95%
Chemical Name or Material Ceftiofur sodium
RTECSNumber XI0367360
TSCA No
storageNote1 2° to 8°C
Recommended Storage Keep cold

RUO - Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.