Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CEL Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA581352
Description
This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). This antibody has specificity for Mouse Bile salt-activated lipase/CEL.
Caboxyl Ester Lipase (CEL), also known as Bile Salt-Activated Lipase, catalyzes the hydrolysis of a wide range of substrates including cholesteryl esters, phospholipids, lysophospholipids, di- and tri-acylglycerols, and fatty acid esters of hydroxy fatty acids (FAHFAs). Preferentially hydrolyzes FAHFAs with the ester bond further away from the carboxylate. Unsaturated FAHFAs are hydrolyzed more quickly than saturated FAHFAs. It also has an essential role in the complete digestion of dietary lipids and their intestinal absorption, along with the absorption of fat-soluble vitamins. [UniProt].
Specifications
CEL | |
Polyclonal | |
Unconjugated | |
CEL | |
1810036E18Rik; BAL; Bile salt-activated lipase; bile salt-dependent lipase, oncofetal isoform; bile salt-stimulated lipase; BSDL; BSSL; bucelipase; carboxyl ester hydrolase; carboxyl ester lipase; carboxyl ester lipase (bile salt-stimulated lipase); CEase; Cel; CELL; Cholesterol esterase; FAP; FAPP; fetoacinar pancreatic protein; Lip1; LIPA; LOW QUALITY PROTEIN: bile salt-activated lipase; lysophospholipase, pancreatic; MODY8; pancreatic lysophospholipase; Sterol esterase | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
12613 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
ELISA, Immunohistochemistry (Paraffin) | |
1 mg/mL | |
PBS with no preservative | |
Q64285 | |
CEL | |
Recombinant Mouse Bile salt-activated lipase/CEL Protein (Met1-Leu534). | |
100 μL | |
Primary | |
Mouse | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction