Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CEL Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA596748
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA596748 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA596748 Supplier Invitrogen™ Supplier No. PA596748
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Samples: Raji, Mouse spleen Immunogen sequence: IDMPAINKGN KKVTEEDFYK LVSEFTITKG LRGAKTTFDV YTESWAQDPS QENKKKTVVD FETDVLFLVP TEIALAQHRA NAKSAKTYAY LFSHPSRMPV YPKWVGADHA DDIQYVFGKP FATPTGYRPQ DRTVSKAMIA YWTNFAKTGD PNMGDSAVPT HWEPYTTENS GYLEITKKMG SSSMKRSLRT NFLRYWTLTY L.

Caboxyl Ester Lipase (CEL), also known as Bile Salt-Activated Lipase, catalyzes the hydrolysis of a wide range of substrates including cholesteryl esters, phospholipids, lysophospholipids, di- and tri-acylglycerols, and fatty acid esters of hydroxy fatty acids (FAHFAs). Preferentially hydrolyzes FAHFAs with the ester bond further away from the carboxylate. Unsaturated FAHFAs are hydrolyzed more quickly than saturated FAHFAs. It also has an essential role in the complete digestion of dietary lipids and their intestinal absorption, along with the absorption of fat-soluble vitamins. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen CEL
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 4.12 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene CEL
Gene Accession No. P07882, P19835, Q64285
Gene Alias 1810036E18Rik; BAL; Bile salt-activated lipase; bile salt-dependent lipase, oncofetal isoform; bile salt-stimulated lipase; BSDL; BSSL; bucelipase; carboxyl ester hydrolase; carboxyl ester lipase; carboxyl ester lipase (bile salt-stimulated lipase); CEase; Cel; CELL; Cholesterol esterase; FAP; FAPP; fetoacinar pancreatic protein; Lip1; LIPA; LOW QUALITY PROTEIN: bile salt-activated lipase; lysophospholipase, pancreatic; MODY8; pancreatic lysophospholipase; Sterol esterase
Gene Symbols CEL
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 350-550 of human CEL (NP_0017982).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1056, 12613, 24254
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.