Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CEL Recombinant Rabbit Monoclonal Antibody (063)
GREENER_CHOICE

Catalog No. PIMA529685
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA529685 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA529685 Supplier Invitrogen™ Supplier No. MA529685
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire. This antibody has specificity for Mouse Bile salt-activated lipase/CEL.

Caboxyl Ester Lipase (CEL), also known as Bile Salt-Activated Lipase, catalyzes the hydrolysis of a wide range of substrates including cholesteryl esters, phospholipids, lysophospholipids, di- and tri-acylglycerols, and fatty acid esters of hydroxy fatty acids (FAHFAs). Preferentially hydrolyzes FAHFAs with the ester bond further away from the carboxylate. Unsaturated FAHFAs are hydrolyzed more quickly than saturated FAHFAs. It also has an essential role in the complete digestion of dietary lipids and their intestinal absorption, along with the absorption of fat-soluble vitamins. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen CEL
Applications Immunohistochemistry (Paraffin)
Classification Recombinant Monoclonal
Clone 063
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene CEL
Gene Accession No. Q64285
Gene Alias 1810036E18Rik; BAL; Bile salt-activated lipase; bile salt-dependent lipase, oncofetal isoform; bile salt-stimulated lipase; BSDL; BSSL; bucelipase; carboxyl ester hydrolase; carboxyl ester lipase; carboxyl ester lipase (bile salt-stimulated lipase); CEase; Cel; CELL; Cholesterol esterase; FAP; FAPP; fetoacinar pancreatic protein; Lip1; LIPA; LOW QUALITY PROTEIN: bile salt-activated lipase; lysophospholipase, pancreatic; MODY8; pancreatic lysophospholipase; Sterol esterase
Gene Symbols CEL
Host Species Rabbit
Immunogen Recombinant Mouse Bile salt-activated lipase/CEL Protein (Met1-Leu534).
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 12613
Target Species Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.