Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CELA1 Antibody, DyLight 594, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP178085DL594
Description
CELA1 Polyclonal antibody specifically detects CELA1 in Mouse, Porcine samples. It is validated for Western Blot, ELISA, ImmunofluorescenceSpecifications
CELA1 | |
Polyclonal | |
Western Blot, ELISA, Immunocytochemistry/Immunofluorescence | |
chymotrypsin-like elastase family member 1, chymotrypsin-like elastase family, member 1, EC 3.4.21, EC 3.4.21.36, ELA1elastase-1, elastase 1, pancreatic, Elastase-1 | |
CELA1 [Porcine Pancreas] (Uniprot: P00772) | |
0.1 mL | |
Cellular Markers, Chromatin Research, Lipid and Metabolism | |
1990 | |
Store at 4C in the dark. |
Western Blot, ELISA, Immunofluorescence | |
DyLight 594 | |
50mM Sodium Borate | |
Rabbit | |
Multi-step | |
RUO | |
Primary | |
Mouse, Pig | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction