Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CELA1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA568395
Description
CELA1 Polyclonal Antibody for IHC (P)
Elastases form a subfamily of serine proteases that hydrolyze many proteins in addition to elastin. Humans have six structurally similar proteins elastase 1, 2, 2A, 2B, 3A, and 3B. Unlike other elastases, pancreatic elastase 1 is not expressed in the pancreas. To date, elastase 1 (ELA1) expression has only been detected in skin keratinocytes. Clinical literature that describes human elastase 1 activity in the pancreas is actually referring to elastase 2A.Elastases form a subfamily of serine proteases that hydrolyze many proteins in addition to elastin. Humans have six elastase genes which encode the structurally similar proteins elastase 1, 2, 2A, 2B, 3A, and 3B. Unlike other elastases, pancreatic elastase 1 is not expressed in the pancreas. To date, elastase 1 expression has only been detected in skin keratinocytes. Clinical literature that describes human elastase 1 activity in the pancreas is actually referring to elastase 2A.
Specifications
CELA1 | |
Polyclonal | |
Unconjugated | |
CELA1 | |
1810009A17Rik; 1810062B19Rik; Cela1; chymotrypsin like elastase 1; chymotrypsin like elastase family member 1; chymotrypsin-like elastase family member 1; chymotrypsin-like elastase family, member 1; ELA1; Ela-1; ELAI1; elastase 1; elastase 1, pancreatic; elastase I; Elastase-1; ELS1; pancreatic elastase 1; pancreatic elastase I; pancreatic preproelastase I; PC-TsF; preproelastase I; RATELAI1 | |
Rabbit | |
affinity chromatography | |
RUO | |
1990 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin) | |
2.2 mg/mL | |
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
Q9UNI1 | |
CELA1 | |
Synthesized peptide derived from human CELA1. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction