Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CELA2A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA571873
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA571873 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA571873 Supplier Invitrogen™ Supplier No. PA571873
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

CELA2A Polyclonal Antibody for Western Blot

Elastases form a subfamily of serine proteases that hydrolyze many proteins in addition to elastin. Humans have six elastase genes which encode the structurally similar proteins elastase 1, 2, 2A, 2B, 3A, and 3B. Like most of the human elastases, elastase 2A is secreted from the pancreas as a zymogen. In other species, elastase 2A has been shown to preferentially cleave proteins after leucine, methionine, and phenylalanine residues.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CELA2A
Applications Western Blot
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene CELA2A
Gene Accession No. P08217
Gene Alias CELA2A; chymotrypsin like elastase family member 2A; chymotrypsin-like elastase family member 2A; chymotrypsin-like elastase family, member 2A; Ela2; Ela-2; Ela2a; ELAII; Elaii1; elastase 2; Elastase 2, pancreatic; elastase 2A; elastase II; Elastase-2; elastase-2A; neutrophilic elastase; pancreatic elastase 2; pancreatic elastase IIA; PE-1; preproelastase II
Gene Symbols CELA2A
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 231-260 amino acids from the C-terminal region of human CELA2A.
Purification Method Antigen Affinity Chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 63036
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.