Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CELA2B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PA549494
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA549494 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA549494 Supplier Invitrogen™ Supplier No. PA549494
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

CELA2B Polyclonal Antibody for Western Blot

Acts upon elastin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CELA2B
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene CELA2B
Gene Accession No. P08218
Gene Alias CELA2B; chymotrypsin like elastase family member 2B; chymotrypsin-like elastase family member 2B; chymotrypsin-like elastase family, member 2B; ELA2B; elastase 2B; elastase-2B; pancreatic elastase IIB (ELA2B); RP11-265F14.2
Gene Symbols CELA2B
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 88-122 amino acids from the Central region of human CELA2B.
Purification Method Antigen Affinity Chromatography, Protein A
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51032
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.