Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CELA3B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5115171
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5115171 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5115171 Supplier Invitrogen™ Supplier No. PA5115171
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total Elastase.

Efficient protease with alanine specificity but only little elastolytic activity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CELA3B
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide
Gene CELA3B
Gene Accession No. P08861, Q9CQ52
Gene Alias 0910001F22Rik; 2310074F01Rik; AI504000; CBPP; Cela3b; cholesterol-binding pancreatic protease; chymotrypsin like elastase family member 3B; chymotrypsin-like elastase family member 3B; chymotrypsin-like elastase family, member 3B; Ela3; ELA3B; elastase 3, pancreatic; elastase 3B, pancreatic; elastase IIIB; Elastase-3B; fecal elastase 1; pancreatic elastase 1; pancreatic endopeptidase E; procarboxypeptidase A complex component III; procarboxypeptidase A-S6 subunit III; PROCPA-S6 III; proproteinase E; protease E; proteinase E
Gene Symbols CELA3B
Host Species Rabbit
Immunogen A synthesized peptide derived from human CELA3B(Accession P08861), corresponding to amino acid residues V68-V118.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23436, 298567, 67868
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.