Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CELA3B Polyclonal Antibody, Invitrogen™
GREENER_CHOICE

Catalog No. FEPA5145363
Change view
Click to view available options
Quantity:
20 μL
200 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEPA5145363 200 μL
FEPA5145364 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. FEPA5145363 Supplier Invitrogen™ Supplier No. PA5145363
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Efficient protease with alanine specificity but only little elastolytic activity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CELA3B
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% Proclin 300; pH 7.4
Gene CELA3B
Gene Accession No. P08861, Q9CQ52
Gene Alias 0910001F22Rik; 2310074F01Rik; AI504000; CBPP; Cela3b; cholesterol-binding pancreatic protease; chymotrypsin like elastase family member 3B; chymotrypsin-like elastase family member 3B; chymotrypsin-like elastase family, member 3B; Ela3; ELA3B; elastase 3, pancreatic; elastase 3B, pancreatic; elastase IIIB; Elastase-3B; fecal elastase 1; pancreatic elastase 1; pancreatic endopeptidase E; procarboxypeptidase A complex component III; procarboxypeptidase A-S6 subunit III; PROCPA-S6 III; proproteinase E; protease E; proteinase E
Gene Symbols CELA3B
Host Species Rabbit
Immunogen Recombinant Human ELA3B protein, Val29-His270 (Accession #P08861).
Purification Method Antigen affinity chromatography, Protein A
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23436, 298567, 67868
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.